Assay Details
Gene Symbol: Yae1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025904.3 | NP_080180.1 | 3 | 3351 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854604 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854599 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854603 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854600 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854602 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963634 | NM_025904.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963634 | AK005425.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963631 | AK151981.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963631 | AK216868.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963631 | AK076034.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963631 | BC048453.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963631 | AK011852.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525638 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963634 | NM_025904.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963634 | AK005425.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963631 | AK151981.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963631 | AK216868.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963631 | AK076034.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963631 | BC048453.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963631 | AK011852.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854608 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854617 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854604 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854594 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854585 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28854589 | - | - |
Whole Mouse Genome G4122A | A_52_P269956 | AK151981.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P269956 | AK076034.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P269956 | BC048453.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P269956 | AK011852.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P269956 | AK019184.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P529098 | AK030455.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309618 | NM_025904.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P309618 | AK005425.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128289 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11106205 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11106204 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11106211 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11106212 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11106208 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854600 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854599 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854603 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854602 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28854604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28854552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854546 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10782337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854593 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10693353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854553 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10453419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854552 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10453416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854609 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10453421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854578 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10820896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854580 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854545 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28854585 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10407919 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497589 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407918 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407920 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10497588 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403758 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403760 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403757 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403759 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449557_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419661_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419660_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10407916 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403756 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10497587 | - | - |
Mouse Genome 430 2.0 Array | 1419661_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419660_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419661_at | - | - |
Mouse Genome 430 2.0 Array | 1449557_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419660_at | - | - |
Mouse Genome 430 2.0 Array | 1449557_at | - | - |
Mouse Expression Set 430 | 1419661_at | - | - |
Mouse Expression Set 430 | 1449557_at | - | - |
Mouse Expression Set 430 | 1419660_PM_at | - | - |
Mouse Expression Set 430 | 1419660_at | - | - |
Mouse Expression Set 430 | 1449557_PM_at | - | - |
Mouse Expression Set 430 | 1419661_PM_at | - | - |
Murine Genome U74v2 Set | 168861_PM_at | - | - |
Murine Genome U74v2 Set | 168861_at | - | - |
Mouse Genome 430A 2.0 Array | 1419660_at | - | - |
Mouse Genome 430A 2.0 Array | 1419661_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449557_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419660_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449557_at | - | - |
Mouse Genome 430A 2.0 Array | 1419661_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449557_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419660_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449557_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419661_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419660_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419661_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 860708 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4120463 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004120463 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683147 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2683147 | - | - |
mouse-6 v1.1 (Illumina) | scl067008.1_17-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4120463 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067008.1_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120463 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683155 | - | - |
