Assay Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK164695.1 | - | 1 | 46 | 109 | |
| BY018870.1 | - | 1 | 44 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164695.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ifi203
Genomic Map