Assay Details
Gene Symbol: Gbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256005.1 | NP_001242934.1 | 11 | 1913 | 99 | |
NM_008620.3 | NP_032646.2 | 11 | 1850 | 99 | ||
GenBank mRNA | AK220567.1 | - | 11 | 1831 | 99 | |
BC141364.1 | - | 11 | 1801 | 9055977 | 99 | |
BC147434.1 | - | 11 | 1801 | 9055976 | 99 | |
EF494423.1 | - | 11 | 1913 | 99 | ||
EF494424.1 | - | 11 | 1884 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
negative regulation of protein phosphorylation
negative regulation of protein ubiquitination
negative regulation of interferon-alpha production
negative regulation of protein import into nucleus
regulation of defense response to virus
cellular response to interferon-gamma
protein binding
negative regulation of transcription from RNA polymerase II promoter
negative regulation of protein phosphorylation
negative regulation of protein ubiquitination
negative regulation of interferon-alpha production
negative regulation of protein import into nucleus
regulation of defense response to virus
cellular response to interferon-gamma
protein binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103837 | EF494423.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103837 | EF494424.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103837 | NM_008620.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2103837 | AK220567.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581963 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408802 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546193 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547548 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44581 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534237 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913173 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811522 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895991 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448987 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67051 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106259 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22616 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104792 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28402 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149481 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103837 | EF494423.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103837 | EF494424.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103837 | NM_008620.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2103837 | AK220567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463562 | BC147434.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463562 | EF494423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463562 | EF494424.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463562 | NM_008620.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463562 | AK220567.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125269 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23954315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954411 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04353914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04125256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04663723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954407 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04425693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04695884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954417 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04547246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23954283 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10532001 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531993 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531988 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532004 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531991 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531992 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532000 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531989 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531998 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532005 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10531990 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532003 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532018 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532009 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532006 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10532016 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10531987 | - | - |
Mouse Genome 430 2.0 Array | 1421104_at | - | - |
Mouse Genome 430 2.0 Array | 1421104_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500025586.mm.1 | TC0500003731.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025586.mm.1 | NM_001256005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025586.mm.1 | NM_001256005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025586.mm.1 | NM_008620.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025586.mm.1 | NM_008620.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025587.mm.1 | TC0500003731.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025587.mm.1 | NM_001256005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025587.mm.1 | NM_001256005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025587.mm.1 | NM_008620.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025587.mm.1 | NM_008620.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025588.mm.1 | TC0500003731.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025588.mm.1 | NM_001256005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025588.mm.1 | NM_001256005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025588.mm.1 | NM_008620.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025588.mm.1 | NM_008620.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025589.mm.1 | TC0500003731.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500025589.mm.1 | NM_001256005.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025589.mm.1 | NM_001256005.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025589.mm.1 | NM_008620.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500025589.mm.1 | NM_008620.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1980670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001980670 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2606792 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2606792 | - | - |
mouse-6 v1.1 (Illumina) | scl0017472.1_81-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1980670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0017472.1_81-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980670 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2606792 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2606792 | - | - |
