Assay Details
| Gene Symbol: Rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277113.1 | NP_001264042.1 | 4 | 2037 | 101 | |
| NM_001277114.1 | NP_001264043.1 | 4 | 2107 | 101 | ||
| NM_009079.3 | NP_033105.1 | 4 | 2087 | 101 | ||
| NR_102291.1 | 5 | 2241 | 101 | |||
| NR_102292.1 | 5 | 2237 | 101 | |||
| GenBank mRNA | AK003454.1 | - | 1 | 728 | 101 | |
| AK009094.1 | - | 4 | 1955 | 101 | ||
| AK012339.1 | - | 1 | 101 | |||
| AK014372.1 | - | 1 | 1221 | 101 | ||
| AK169078.1 | - | 4 | 1940 | 101 | ||
| AK169731.1 | - | 4 | 1953 | 101 | ||
| AK214896.1 | - | 1 | 134 | 101 | ||
| BC021344.1 | - | 4 | 1946 | 4190569 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl22
Genomic Map