Assay Details
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272097.1 | NP_001259026.1 | 4 | 666 | 92 | |
NM_001272098.1 | NP_001259027.1 | 3 | 496 | 92 | ||
NM_010634.3 | NP_034764.1 | 4 | 669 | 92 | ||
GenBank mRNA | AK008782.1 | - | 4 | 663 | 92 | |
AK011551.1 | - | 4 | 665 | 92 | ||
BE987062.1 | - | 1 | 92 | |||
X70100.1 | - | 4 | 648 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
