Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700000260.mm.1 |
TC0700000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000260.mm.1 |
NM_029767.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000260.mm.1 |
NM_029767.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000259.mm.1 |
TC0700000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000259.mm.1 |
NM_029767.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000259.mm.1 |
NM_029767.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000261.mm.1 |
TC0700000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000261.mm.1 |
NM_029767.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000261.mm.1 |
NM_029767.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000258.mm.1 |
TC0700000019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700000258.mm.1 |
NM_029767.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700000258.mm.1 |
NM_029767.2 |
Refseq |