Assay Details
| Gene Symbol: Eif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011508.1 | NP_035638.1 | 4 | 1142 | 80 | |
| GenBank mRNA | AF129888.1 | - | 4 | 1131 | 80 | |
| AK151070.1 | - | 4 | 1139 | 80 | ||
| BC003463.1 | - | 4 | 1142 | 2648657 | 80 | |
| BC081429.1 | - | 4 | 1129 | 5710951 | 80 | |
| BC096656.1 | - | 4 | 1117 | 5143414 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067421 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067417 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067418 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126425 | BC003463.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126425 | BC081429.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126425 | BC096656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126425 | NM_011508.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P126425 | AF129888.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281441 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126425 | BC003463.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126425 | BC081429.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126425 | BC096656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126425 | NM_011508.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P126425 | AF129888.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28067406 | - | - |
| Whole Mouse Genome G4122A | A_52_P74044 | - | - |
| Whole Mouse Genome G4122A | A_51_P283837 | AF129888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283837 | NM_001110020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283837 | BC134385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283837 | Z50159.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P283837 | AK169763.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126425 | BC003463.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126425 | BC081429.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126425 | BC096656.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126425 | NM_011508.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126425 | AF129888.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09552903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09552909 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09552930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09552914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067421 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067423 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067418 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28067417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28067438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09056764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09727672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09474819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09144983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09608566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09406921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09727673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09552909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09028629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28067403 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10564811 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381117 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423799_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423798_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10564809 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597573 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381115 | - | - |
| Mouse Genome 430 2.0 Array | 1423799_at | - | - |
| Mouse Genome 430 2.0 Array | 1423798_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423799_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423798_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20918_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20918_PM_at | - | - |
| Mouse Expression Set 430 | 1423799_at | - | - |
| Mouse Expression Set 430 | 1423798_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423799_PM_at | - | - |
| Mouse Expression Set 430 | 1423798_a_at | - | - |
| Murine Genome U74v2 Set | 92855_at | - | - |
| Murine Genome U74v2 Set | 92855_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423798_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423799_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423798_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423799_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423798_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423798_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423799_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423799_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4570471 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4570471 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570471 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213600 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213600 | - | - |
| mouse-6 v1.1 (Illumina) | scl020918.4_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570471 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020918.4_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570471 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2731940 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl020918.4_11-S | - | - |

Eif1
Genomic Map