Assay Details
Gene Symbol: Txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172054.4 | NP_742051.1 | 5 | 4048 | 82 | |
GenBank mRNA | AK153857.1 | - | 5 | 4049 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157010 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157016 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157015 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157018 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157012 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157014 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157019 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162552 | BC083077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162552 | AK028525.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162552 | AK153857.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162552 | AK167045.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162552 | AK002893.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1979221 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162557 | AK153857.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162557 | NM_172054.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616686 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911282 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574474 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784982 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525875 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482193 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16024 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9808 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132498 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175432 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1979221 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162557 | AK153857.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162557 | NM_172054.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162552 | BC083077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162552 | AK028525.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162552 | AK153857.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162552 | AK167045.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162552 | AK002893.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20157019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20156993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04067463 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132498 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175432 | - | - |
Whole Mouse Genome G4122A | A_51_P108723 | AK028525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P108723 | AK153857.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P108723 | AK002893.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P108723 | NM_172054.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P345777 | BC083077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P345777 | AK028525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P345777 | AK153857.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P345777 | AK167045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P345777 | BC022947.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P253136 | AK028525.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P253136 | AK085023.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P253136 | AK153857.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P253136 | NM_172054.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127167 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136740 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175432 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132498 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00957894 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00957886 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00957881 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00957871 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157010 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157018 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157019 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157012 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157014 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157015 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20157016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20157019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20156956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00698483 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20157000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00170558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00644924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00146276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00146280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01009862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01009861 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20156984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00698480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00957893 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10396065 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10354081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10354084 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10354080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10354082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10354083 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424512_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436108_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436951_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10396064 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10354079 | - | - |
Mouse Genome 430 2.0 Array | 1424512_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1436108_at | - | - |
Mouse Genome 430 2.0 Array | 1436951_x_at | - | - |
Mouse Genome 430 2.0 Array | 1436108_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424512_a_at | - | - |
Mouse Genome 430 2.0 Array | 1436951_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98258_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98258_PM_at | - | - |
Mouse Expression Set 430 | 1424512_a_at | - | - |
Mouse Expression Set 430 | 1424512_PM_a_at | - | - |
Mouse Expression Set 430 | 1436951_PM_x_at | - | - |
Mouse Expression Set 430 | 1436951_x_at | - | - |
Mouse Expression Set 430 | 1436108_at | - | - |
Mouse Expression Set 430 | 1436108_PM_at | - | - |
Murine Genome U74v2 Set | 97900_PM_at | - | - |
Murine Genome U74v2 Set | 97900_at | - | - |
Murine Genome U74v2 Set | 166178_PM_at | - | - |
Murine Genome U74v2 Set | 166178_at | - | - |
Mouse Genome 430A 2.0 Array | 1424512_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1424512_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1436951_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1436951_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424512_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424512_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436951_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436951_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436108_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436108_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1580079 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1580079 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001580079 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255365 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255365 | - | - |
mouse-6 v1.1 (Illumina) | scl16848.5_127-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1580079 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl16848.5_127-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580079 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1255365 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255365 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl16848.5_127-S | - | - |
