Assay Details
Gene Symbol: Khdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183322.2 | NP_899145.2 | 3 | 1851 | 116 | |
GenBank mRNA | AK162365.1 | - | 1 | 947 | 116 | |
AY238603.1 | - | 3 | 1614 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | AK162365.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | AY238603.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | AK162365.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | AY238603.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | AK162365.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | AY238603.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | AK162365.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | AY238603.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077466 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20077470 | - | - |
Whole Mouse Genome G4122A | A_52_P208521 | AK162365.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P208521 | AY238603.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P208521 | AK162365.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P208521 | AY238603.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P208521 | NM_183322.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | BC099381.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | BC141037.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | NM_001033904.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | AK136166.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | AY238603.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | BC099381.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | BC141037.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | NM_001033904.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | AK136166.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P70949 | AY238603.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20077450 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00307535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01038385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00572799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01091028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01091029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00377129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00099836 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077451 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077523 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00377130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00608934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077485 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00099835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20077462 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10345078 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345084 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10345085 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455423_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10345077 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10345083 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10345079 | - | - |
Mouse Genome 430 2.0 Array | 1455423_at | - | - |
Mouse Genome 430 2.0 Array | 1455423_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 368204_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 368204_at | - | - |
Mouse Expression Set 430 | 1455423_at | - | - |
Mouse Expression Set 430 | 1455423_PM_at | - | - |
Murine Genome U74v2 Set | 103992_f_at | - | - |
Murine Genome U74v2 Set | 103992_PM_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1455423_at | - | - |
Mouse Genome 430A 2.0 Array | 1455423_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455423_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1455423_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 391820 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 460563 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 460563 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000460563 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612439 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2612439 | - | - |
mouse-6 v1.1 (Illumina) | scl0368204.3_330-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 460563 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0368204.3_330-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460563 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612439 | - | - |
