Assay Details
| Gene Symbol: Defb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013756.2 | NP_038784.1 | 2 | 152 | 108 | |
| GenBank mRNA | AF092929.1 | - | 2 | 149 | 108 | |
| BC114344.2 | - | 2 | 149 | 40049236 | 108 | |
| BC116176.2 | - | 2 | 153 | 40049224 | 108 | |
| BC116177.1 | - | 2 | 152 | 40049232 | 108 | |
| BC134358.1 | - | 2 | 153 | 40049234 | 108 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116206 | NM_013756.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116206 | AF092929.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62048 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111642 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116206 | NM_013756.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116206 | AF092929.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25702823 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111642 | - | - |
| Whole Mouse Genome G4122A | A_51_P357996 | BC116177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P357996 | BC134358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P357996 | AF092929.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P357996 | BC114344.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P357996 | BC116176.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116206 | NM_013756.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116206 | AF092929.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111642 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07364760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25702877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07418082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06976155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06868525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06868526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07418090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06885437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07438054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25702822 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570623 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421806_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570621 | - | - |
| Mouse Genome 430 2.0 Array | 1421806_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421806_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27358_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27358_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001614.mm.1 | NM_013756.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001614.mm.1 | NM_013756.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001614.mm.1 | TC0800000185.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421806_at | - | - |
| Mouse Expression Set 430 | 1421806_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421806_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421806_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421806_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421806_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 411101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 50014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000050014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610322 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610322 | - | - |
| mouse-6 v1.1 (Illumina) | scl34016.2.1_50-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 50014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34016.2.1_50-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50014 | - | - |

Defb3
Genomic Map