Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200003158.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003158.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003158.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003155.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003155.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003155.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003151.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003151.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003151.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003150.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003150.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003150.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003153.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003153.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003153.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003154.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003154.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003154.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003149.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003149.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003149.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003156.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003156.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003156.mm.1 |
TC1200000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200003157.mm.1 |
NM_001100109.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003157.mm.1 |
NM_001100109.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200003157.mm.1 |
TC1200000460.mm.1 |
MTA TC |