| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10584253 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10584252 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900002573.mm.1 |
NM_001128510.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002573.mm.1 |
NM_001128510.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002573.mm.1 |
TC0900000345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002572.mm.1 |
NM_001128510.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002572.mm.1 |
NM_001128510.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002572.mm.1 |
TC0900000345.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002574.mm.1 |
NM_001128510.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002574.mm.1 |
NM_001128510.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002574.mm.1 |
TC0900000345.mm.1 |
MTA TC |