Assay Details
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110555.1 | NP_001104025.1 | 2 | 1754 | 67 | |
GenBank mRNA | BC011440.1 | - | 3 | 1829 | 3989862 | 67 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097979.2 | NP_001091448.2 | 2 | 1754 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
