Assay Details
| Gene Symbol: Hist1h2bq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097979.2 | NP_001091448.2 | 2 | 1847 | 112 | |
| Gene Symbol: Hist1h2br | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110555.1 | NP_001104025.1 | 2 | 1847 | 112 | |
| GenBank mRNA | BC011440.1 | - | 3 | 1920 | 3989862 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | BC011440.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | NM_001110555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559748 | NM_001097979.2 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10403979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408211 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10403979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408091 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403978 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403959 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408087 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403980 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403980 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403978 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408087 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408210 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408070 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408070 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408210 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10403959 | - | - |
| Mouse Genome 430 2.0 Array | 1425398_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425398_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001213.mm.1 | NM_001110555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001213.mm.1 | NM_001110555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001213.mm.1 | TC1300000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001212.mm.1 | NM_001110555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001212.mm.1 | NM_001110555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001212.mm.1 | TC1300000183.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009855.mm.1 | NM_001097979.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009855.mm.1 | NM_001097979.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009855.mm.1 | TC1300001563.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009854.mm.1 | NM_001097979.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009854.mm.1 | NM_001097979.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009854.mm.1 | TC1300001563.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009858.mm.1 | NM_001097979.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009858.mm.1 | NM_001097979.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009858.mm.1 | TC1300001563.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001208.mm.1 | NM_001110555.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001208.mm.1 | NM_001110555.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001208.mm.1 | TC1300000183.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425398_PM_at | - | - |
| Mouse Expression Set 430 | 1452540_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452540_PM_a_at | - | - |
| Mouse Expression Set 430 | 1418072_PM_at | - | - |
| Mouse Expression Set 430 | 1418072_PM_at | - | - |
| Mouse Expression Set 430 | 1452540_a_at | - | - |
| Mouse Expression Set 430 | 1452540_a_at | - | - |
| Mouse Expression Set 430 | 1418072_at | - | - |
| Mouse Expression Set 430 | 1418072_at | - | - |
| Mouse Expression Set 430 | 1425398_PM_at | - | - |
| Mouse Expression Set 430 | 1425398_at | - | - |
| Mouse Expression Set 430 | 1425398_at | - | - |
| Murine Genome U74v2 Set | 101654_PM_at | - | - |
| Murine Genome U74v2 Set | 93833_s_at | - | - |
| Murine Genome U74v2 Set | 101654_at | - | - |
| Murine Genome U74v2 Set | 93833_PM_s_at | - | - |
| Murine Genome U74v2 Set | 101654_PM_at | - | - |
| Murine Genome U74v2 Set | 101654_at | - | - |
| Murine Genome U74v2 Set | 93833_PM_s_at | - | - |
| Murine Genome U74v2 Set | 93833_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418072_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425398_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418072_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425398_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418072_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452540_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452540_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452540_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452540_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425398_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418072_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425398_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 601312 | - | - |
| ABI Mouse Genome Survey Microarray | 899211 | - | - |

Hist1h2bq
Genomic Map