Assay Details
| Gene Symbol: Mirlet7g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029526.1 | 1 | 47 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycerate kinase | Glyctk |
6230410P16Rik, C430006L12 |
-17.8kb | Anti-sense | Chr. 9 - 106152860 - 106161065 on NCBI Build GRCm38 |
| Downstream | protein phosphatase 1M | Ppm1m |
2810423O19Rik, AW610647, C77250, PP2Ceta |
16.0kb | Anti-sense | Chr. 9 - 106194953 - 106199233 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7g
Genomic Map