Assay Details
Gene Symbol: Gm2762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037991.1 | 2 - 3 | 174 | 103 | ||
GenBank mRNA | AK006343.1 | - | 2 - 3 | 168 | 103 | |
AK030238.1 | - | 2 - 3 | 175 | 103 | ||
BB617179.1 | - | 2 - 3 | 175 | 103 | ||
BC100495.1 | - | 2 - 3 | 155 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
serine palmitoyltransferase, long chain base subunit 1 |
Sptlc1 |
AW552086, C77762, E030036H05, Lcb1 |
-16.8kb | Anti-sense | Chr. 13 - 53332748 - 53377417 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700058M13 gene | 1700058M13Rik | 2.7kb | Anti-sense | Chr. 13 - 53398086 - 53415880 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
