Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308247 |
AK034617.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308247 |
AK139382.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308247 |
BC024059.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
308247 |
NM_201361.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121715 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510793 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
572480 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569091 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
87738 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
754762 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
477071 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
728674 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
512075 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
929274 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
676156 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
101115 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10246 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994962 |
NR_038008.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1994962 |
AK138346.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04241589 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00169836 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04241589 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565183 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565184 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565165 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565173 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565175 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565168 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01127934 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565177 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565196 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00589956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565167 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01012252 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565193 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565204 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565194 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565182 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565170 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565185 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565188 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565200 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565190 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00226830 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00865807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565164 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565198 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20565191 |
- |
- |