Assay Details
| Gene Symbol: 4930481A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027938.2 | 3 - 4 | 551 | 91 | ||
| GenBank mRNA | BM937611.1 | - | 3 - 4 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific 10 interacting protein | Tsga10ip |
1810013N20Rik, Fam161c |
-14.9kb | Anti-sense | Chr. 19 - 5390047 - 5402534 on NCBI Build GRCm38 |
| Downstream | expressed sequence AI837181 | AI837181 |
Bles03, N28173 |
2.3kb | Sense | Chr. 19 - 5425144 - 5427317 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930481A15Rik
Genomic Map