Assay Details
| Gene Symbol: Snord83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028282.1 | 1 | 34 | 64 | ||
| GenBank mRNA | AF357381.1 | - | 1 | 34 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L3 | Rpl3 |
F2, J1 |
Sense | Chr. 15 - 80077781 - 80083406 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, C/D box 43 |
Snord43 | MBII-47 | 4.3kb | Sense | Chr. 15 - 80082859 - 80082906 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord83b
Genomic Map