Assay Details
| Gene Symbol: C4bp-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028304.1 | 5 - 6 | 547 | 70 | ||
| GenBank mRNA | BB728650.1 | - | 2 - 3 | 325 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | complement component 4 binding protein | C4bp |
AI195242, C4bpa |
-8.6kb | Sense | Chr. 1 - 130635921 - 130661618 on NCBI Build GRCm38 |
| Downstream | 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 | Pfkfb2 | 4930568D07Rik | 18.0kb | Sense | Chr. 1 - 130689043 - 130729253 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C4bp-ps1
Genomic Map