Assay Details
| Gene Symbol: Snord71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028532.1 | 1 | 34 | 64 | ||
| GenBank mRNA | AF357330.1 | - | 1 | 34 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ataxin 1-like | Atxn1l |
AW558552, Boat, C330011L24Rik, D6Mgi37, D8Ertd587e, ENSMUSG00000069895 |
-101.5kb | Anti-sense | Chr. 8 - 109726451 - 109737796 on NCBI Build GRCm38 |
| Downstream | PH domain and leucine rich repeat protein phosphatase 2 | Phlpp2 |
AI481772, C130044A18Rik, Phlppl |
29.1kb | Sense | Chr. 8 - 109868519 - 109944671 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snord71
Genomic Map