Assay Details
| Gene Symbol: Gm10863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029470.1 | 3 - 4 | 329 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SRY (sex determining region Y)-box 10 | Sox10 |
Dom, Sox21 |
-44.7kb | Anti-sense | Chr. 15 - 79154909 - 79165235 on NCBI Build GRCm38 |
| Downstream | protein interacting with C kinase 1 | Pick1 | Prkcabp | 18.9kb | Sense | Chr. 15 - 79229166 - 79249466 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm10863
Genomic Map