Assay Details
| Gene Symbol: Mir30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029533.1 | 1 | 36 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 29506 | Gm29506 | -36.6kb | Anti-sense | Chr. 1 - 23228493 - 23235658 on NCBI Build GRCm38 | |
| Downstream | microRNA 30c-2 | Mir30c-2 |
Mirn30c-2, mir-30c-2 |
19.4kb | Sense | Chr. 1 - 23291701 - 23291784 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir30a
Genomic Map