Assay Details
| Gene Symbol: Mir30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029534.1 | 1 | 53 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and AT hook domain containing | Zfat |
Gm922, Zfat1, Zfp406 |
-78.2kb | Sense | Chr. 15 - 68083714 - 68259265 on NCBI Build GRCm38 |
| Downstream | microRNA 30d | Mir30d |
Mirn30d, mir-30d, mmu-mir-30d |
3.7kb | Sense | Chr. 15 - 68341208 - 68341289 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir30b
Genomic Map