Assay Details
| Gene Symbol: Mir99a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029535.1 | 1 | 33 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31322 |
Gm31322 | Sense | Chr. 16 - 77589379 - 77790662 on NCBI Build GRCm38 | ||
| Downstream | microRNA let7c-1 | Mirlet7c-1 |
Mirnlet7c, Mirnlet7c-1, let-7c-1 |
657.0 | Sense | Chr. 16 - 77599657 - 77599750 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir99a
Genomic Map