Assay Details
| Gene Symbol: Mir195a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029581.1 | 1 | 43 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 497b | Mir497b |
Gm27737, mmu-mir-497b |
-226.0 | Anti-sense | Chr. 11 - 70234692 - 70234816 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 0610010K14 gene | 0610010K14Rik |
1110020A23Rik, AL033328, AU045833, Bap18 |
69.0 | Anti-sense | Chr. 11 - 70235204 - 70237914 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir195a
Genomic Map