Assay Details
| Gene Symbol: Mir322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029756.1 | 1 | 53 | 60 | ||
| Gene Symbol: C430049B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038184.1 | 4 | 1808 | 60 | ||
| NR_038185.1 | 4 | 1522 | 60 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 503 | Mir503 |
Mirn503, mmu-mir-503 |
-201.0 | Sense | Chr. X - 53053984 - 53054054 on NCBI Build GRCm38 |
| Downstream | placental specific protein 1 | Plac1 |
DXWsu72e, Epcs26, Oosp2l |
15.6kb | Sense | Chr. X - 53069993 - 53240122 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir322
Genomic Map