Assay Details
| Gene Symbol: Mir330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029763.1 | 1 | 50 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gastric inhibitory polypeptide receptor | Gipr |
GIP-R, Gm1081, Gm160 |
-11.2kb | Anti-sense | Chr. 7 - 19156063 - 19170248 on NCBI Build GRCm38 |
| Downstream | G protein-coupled receptor 4 | Gpr4 | 30.9kb | Sense | Chr. 7 - 19212499 - 19224176 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir330
Genomic Map