Assay Details
| Gene Symbol: Mir107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029783.1 | 1 | 46 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 16 (monocarboxylic acid transporters), member 12 | Slc16a12 | 3230401A21, AW210596 | -72.4kb | Sense | Chr. 19 - 34668401 - 34748269 on NCBI Build GRCm38 | 
| Downstream | kinesin family member 20B | Kif20b | 33cex, B130024C23, C330014J10Rik, Mphosph1, magoo | 101.6kb | Anti-sense | Chr. 19 - 34922338 - 34975750 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length between 71 and 85  3' Most  Detect Genomic DNA  Mus musculus  Both primers and probe map within a single exon | 
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Mir107
Mir107 Genomic Map
Genomic Map Back To Top
 Back To Top