Assay Details
| Gene Symbol: Mir17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029785.1 | 1 | 35 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700100I10 gene | 1700100I10Rik |
1700085D13Rik, 4930481K08Rik |
-97.7kb | Anti-sense | Chr. 14 - 114940225 - 114945948 on NCBI Build GRCm38 |
| Downstream | microRNA 18 | Mir18 |
Mirn18, mir-18a, mmu-mir-18, mmu-mir-18a |
97.0 | Sense | Chr. 14 - 115043851 - 115043946 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir17
Genomic Map