Assay Details
| Gene Symbol: Mir181a-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029795.1 | 1 | 35 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4258 | Gm4258 | -5.7kb | Sense | Chr. 1 - 137898476 - 137960785 on NCBI Build GRCm38 | |
| Downstream | microRNA 181b-1 | Mir181b-1 |
Mirn181b-1, mir-181b-1 |
98.0 | Sense | Chr. 1 - 137966639 - 137966718 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir181a-1
Genomic Map