Assay Details
| Gene Symbol: Mir19b-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029815.1 | 1 | 42 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 20a | Mir20a |
Mir-20, Mirn20, Mirn20a, mir-20a, mmu-mir-20a |
-42.0 | Sense | Chr. 14 - 115044157 - 115044263 on NCBI Build GRCm38 |
| Downstream | microRNA 92-1 | Mir92-1 |
Mirn92-1, mir-92a-1 |
36.0 | Sense | Chr. 14 - 115044427 - 115044506 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir19b-1
Genomic Map