Assay Details
| Gene Symbol: Mir7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029827.1 | 1 | 50 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A230051N06 gene | A230051N06Rik | -183.0 | Sense | Chr. 17 - 56241717 - 56242805 on NCBI Build GRCm38 | |
| Downstream | feminization 1 homolog a (C. elegans) | Fem1a |
AW611390, Fem1aa |
13.7kb | Sense | Chr. 17 - 56256793 - 56263608 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7b
Genomic Map