Assay Details
| Gene Symbol: Mir452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029974.1 | 1 | 43 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 224 | Mir224 |
Mirn224, mir-224, mmu-mir-224 |
-1.1kb | Sense | Chr. X - 72261031 - 72261112 on NCBI Build GRCm38 |
| Downstream |
melanoma antigen family A, 10 |
Magea10 | 119.6kb | Sense | Chr. X - 72381870 - 72386858 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir452
Genomic Map