Assay Details
| Gene Symbol: Mir540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030260.1 | 1 | 34 | 56 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 337 | Mir337 |
Mirn337, mir-337, mmu-mir-337 |
-195.0 | Sense | Chr. 12 - 109585789 - 109585885 on NCBI Build GRCm38 |
| Downstream | microRNA 665 | Mir665 |
Mirn665, mir-665, mmu-mir-665 |
168.0 | Sense | Chr. 12 - 109586314 - 109586407 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir540
Genomic Map