Assay Details
Gene Symbol: Mir744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030417.1 | 1 | 42 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitogen-activated protein kinase kinase 4 | Map2k4 |
JNKK1, MEK4, MKK4, PRKMK4, Sek1, Serk1 |
Sense | Chr. 11 - 65688244 - 65788354 on NCBI Build GRCm38 | |
Downstream | zinc finger with KRAB and SCAN domains 6 | Zkscan6 |
1700128E15Rik, D11Ertd714e, KOX11, Zfp535, Znf18 |
72.3kb | Anti-sense | Chr. 11 - 65807175 - 65829239 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
