Assay Details
Gene Symbol: Mir763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030434.1 | 1 | 62 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | high mobility group AT-hook 2 | Hmga2 |
9430083A20Rik, HMGI-C, Hmgic, pg, pygmy |
Sense | Chr. 10 - 120361275 - 120476935 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4921513I03 gene | 4921513I03Rik | 317.6kb | Anti-sense | Chr. 10 - 120765732 - 120778886 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
