Assay Details
| Gene Symbol: Mir708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030489.1 | 1 | 55 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | frataxin pseudogene | Gm5037 |
EG260347, Frda-ps |
-336.0kb | Anti-sense | Chr. 7 - 95912745 - 95913390 on NCBI Build GRCm38 |
| Downstream | microRNA 6394 | Mir6394 |
Gm27929, mmu-mir-6394 |
56.1kb | Sense | Chr. 7 - 96305667 - 96305765 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir708
Genomic Map