Assay Details
| Gene Symbol: Mir711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030492.1 | 1 | 42 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquinol-cytochrome c reductase core protein 1 | Uqcrc1 | 1110032G10Rik | -20.3kb | Sense | Chr. 9 - 108936648 - 108949641 on NCBI Build GRCm38 |
| Downstream | urocortin 2 | Ucn2 |
Ucn II, Ucn-2, ucn-II |
16.2kb | Sense | Chr. 9 - 108986163 - 108987164 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir711
Genomic Map