Assay Details
| Gene Symbol: Mir193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030549.1 | 1 | 45 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 41421 |
Gm41421 | -9.3kb | Sense | Chr. 16 - 13436491 - 13440201 on NCBI Build GRCm38 | |
| Downstream | microRNA 365-1 | Mir365-1 |
Mirn365-1, mir-365-1 |
4.2kb | Sense | Chr. 16 - 13453840 - 13453926 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir193b
Genomic Map