Assay Details
| Gene Symbol: Mir875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030606.1 | 1 | 34 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 599 | Mir599 | mmu-mir-599 | -52.0 | Sense | Chr. 15 - 35660831 - 35660918 on NCBI Build GRCm38 |
| Downstream | cytochrome c oxidase subunit VIc | Cox6c | 270.9kb | Sense | Chr. 15 - 35931976 - 35938246 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir875
Genomic Map