Assay Details
| Gene Symbol: Gm15760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030670.1 | 1 | 56 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, sub-family F (GCN20), member 3 |
Abcf3 |
AI326318, AU016058, BB119416 |
Anti-sense | Chr. 16 - 20548556 - 20561603 on NCBI Build GRCm38 | |
| Downstream | von Willebrand factor A domain containing 5B2 | Vwa5b2 |
BB182430, EG328644, Gm1738, Gm931 |
40.8kb | Anti-sense | Chr. 16 - 20589319 - 20605378 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15760
Genomic Map