Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500008703.mm.1 |
TC0500001029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008703.mm.1 |
NR_030673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008703.mm.1 |
NR_030673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008705.mm.1 |
TC0500001029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008705.mm.1 |
NR_030673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008705.mm.1 |
NR_030673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008702.mm.1 |
TC0500001029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008702.mm.1 |
NR_030673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008702.mm.1 |
NR_030673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008701.mm.1 |
TC0500001029.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500008701.mm.1 |
NR_030673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500008701.mm.1 |
NR_030673.1 |
Refseq |