Assay Details
| Gene Symbol: D930016D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030673.1 | 3 - 4 | 482 | 97 | ||
| GenBank mRNA | BC060616.1 | - | 3 - 4 | 449 | 97 | |
| CN457361.1 | - | 3 - 4 | 480 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC005561 | BC005561 | -18.8kb | Sense | Chr. 5 - 104508352 - 104522383 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 33B | Zfp33b | 2310012P17Rik | 16.7kb | Sense | Chr. 5 - 104570873 - 104584620 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136737 | BC060616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136737 | BC082597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136737 | AK140900.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136737 | AK086246.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136737 | AK040297.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179434 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680600 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157979 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680211 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177632 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680603 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680260 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680317 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680325 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680520 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177892 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680382 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680396 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680479 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162801 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680798 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177793 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681138 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177979 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680651 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178283 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103947 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680686 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100729 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177987 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178113 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177694 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680510 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680232 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680244 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680014 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05679919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01177510 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680455 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680492 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178002 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01178072 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05679897 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680417 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680134 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05681152 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05680773 | - | - |
| Whole Mouse Genome G4122A | A_52_P14342 | BC060616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14342 | BC082597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14342 | AK140900.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14342 | AK086246.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P14342 | NR_030673.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27725 | BC023132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27725 | BC060616.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27725 | BC082597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27725 | AK081399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P27725 | NR_030673.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179434 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01177632 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146805 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110135 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157979 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121278 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680396 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103947 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162801 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01178021 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680761 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01177987 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680244 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680520 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680510 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100729 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05680317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04390885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04950813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04531736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04920129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04920128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04258027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23952749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04971455 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523751 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424861_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424860_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523750 | - | - |
| Mouse Genome 430 2.0 Array | 1424860_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424861_at | - | - |
| Mouse Genome 430 2.0 Array | 1424861_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424860_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500008718.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008718.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008718.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008719.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008719.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008719.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008723.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008723.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008723.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008727.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008727.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008727.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008730.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008730.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008730.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008731.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008731.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008731.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008733.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008733.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008733.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008734.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008734.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008734.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008725.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008725.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008725.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008726.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008726.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008726.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008735.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008735.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008735.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008722.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008722.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008722.mm.1 | NR_030673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008724.mm.1 | TC0500001029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008724.mm.1 | NR_030673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008724.mm.1 | NR_030673.1 | Refseq |
| Affymetrix HT Mouse Genome 430A Array | 1424861_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424861_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424860_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424860_PM_at | - | - |

D930016D06Rik
Genomic Map