Assay Details
| Gene Symbol: 2810410L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030682.1 | 1 - 2 | 416 | 93 | ||
| GenBank mRNA | AK164256.1 | - | 1 - 2 | 417 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 9734 | Gm9734 | ENSMUSG00000056649 | -924.0 | Anti-sense | Chr. 11 - 120181536 - 120187565 on NCBI Build GRCm38 |
| Downstream | predicted gene 11770 | Gm11770 | OTTMUSG00000004111 | 23.2kb | Sense | Chr. 11 - 120213034 - 120222100 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK162474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK164256.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | NR_030682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149407 | AK013071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK081626.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK162474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | NR_030682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083789 | AK013071.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882515 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK162474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK164256.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | NR_030682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149407 | AK013071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK081626.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK162474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | NR_030682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083789 | AK013071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P626035 | AK081626.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P626035 | AK038227.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59285 | AK162474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P59285 | AK013071.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09185606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09580839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09185603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09198963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09124925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09328509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28183026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28182968 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1429510_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438972_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393749 | - | - |
| Mouse Genome 430 2.0 Array | 1429510_at | - | - |
| Mouse Genome 430 2.0 Array | 1429510_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438972_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438972_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042332_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100042332_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100039986.mm.1 | NR_030682.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039986.mm.1 | NR_030682.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039986.mm.1 | TC1100004198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039974.mm.1 | NR_030682.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039974.mm.1 | NR_030682.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039974.mm.1 | TC1100004198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039977.mm.1 | NR_030682.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039977.mm.1 | NR_030682.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039977.mm.1 | TC1100004198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039982.mm.1 | NR_030682.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039982.mm.1 | NR_030682.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039982.mm.1 | TC1100004198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100039975.mm.1 | NR_030682.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039975.mm.1 | NR_030682.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100039975.mm.1 | TC1100004198.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438972_x_at | - | - |
| Mouse Expression Set 430 | 1429510_at | - | - |
| Mouse Expression Set 430 | 1438972_PM_x_at | - | - |
| Mouse Expression Set 430 | 1429510_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438972_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429510_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438972_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429510_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 520722 | - | - |

2810410L24Rik
Genomic Map