Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400021679.mm.1 |
TC0400002668.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021679.mm.1 |
NR_033140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021679.mm.1 |
NR_033140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021675.mm.1 |
TC0400002668.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021675.mm.1 |
NR_033139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021675.mm.1 |
NR_033139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021675.mm.1 |
NR_033140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021675.mm.1 |
NR_033140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021674.mm.1 |
TC0400002668.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021674.mm.1 |
NR_033139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021674.mm.1 |
NR_033139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021674.mm.1 |
NR_033140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021674.mm.1 |
NR_033140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021673.mm.1 |
TC0400002668.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021673.mm.1 |
NR_033139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021673.mm.1 |
NR_033139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021673.mm.1 |
NR_033140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021673.mm.1 |
NR_033140.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021676.mm.1 |
TC0400002668.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021676.mm.1 |
NR_033139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021676.mm.1 |
NR_033139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021676.mm.1 |
NR_033140.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021676.mm.1 |
NR_033140.1 |
Refseq |