Assay Details
| Gene Symbol: BC031361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033221.1 | 1 | 745 | 84 | ||
| GenBank mRNA | BC031361.1 | - | 1 | 277 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glycogen synthase kinase 3 beta | Gsk3b |
7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3 |
Anti-sense | Chr. 16 - 38089001 - 38246080 on NCBI Build GRCm38 | |
| Downstream |
nuclear receptor subfamily 1, group I, member 2 |
Nr1i2 |
PXR, PXR.1, PXR.2, PXR1, SXR, mPXR |
159.1kb | Sense | Chr. 16 - 38248321 - 38294851 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BC031361
Genomic Map