Assay Details
| Gene Symbol: Gm10494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033462.1 | 2 - 3 | 505 | 82 | ||
| GenBank mRNA | AK135441.1 | - | 2 - 3 | 506 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930429F11 gene | 4930429F11Rik | -31.3kb | Sense | Chr. 17 - 79475624 - 79477885 on NCBI Build GRCm38 | |
| Downstream | regulator of microtubule dynamics 2 | Rmdn2 |
AW061290, Fam82a1, RMD-2, mRMD-2 |
68.7kb | Anti-sense | Chr. 17 - 79578598 - 79685067 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135441.1 transcripts.
Set Membership: |
|

Gm10494
Genomic Map