Assay Details
Gene Symbol: Gm10823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033475.1 | 2 - 3 | 162 | 84 | ||
GenBank mRNA | AK136620.1 | - | 2 - 3 | 163 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 50 | Ccdc50 | C3orf6 | -447.5kb | Sense | Chr. 16 - 27388765 - 27452218 on NCBI Build GRCm38 |
Downstream | fibroblast growth factor 12 | Fgf12 |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
246.1kb | Anti-sense | Chr. 16 - 28157783 - 28753243 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000878.rn
Additional Information:
This assay is in the 5'UTR region of the AK136620.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
