Assay Details
Gene Symbol: Gm14164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033505.1 | 3 - 4 | 300 | 88 | ||
GenBank mRNA | AY512911.1 | - | 3 - 4 | 300 | 88 | |
AY512942.1 | - | 3 - 4 | 300 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neurensin 2 | Nrsn2 |
Gm123, Neurensin-2 |
Anti-sense | Chr. 2 - 152368755 - 152377153 on NCBI Build GRCm38 | |
Downstream | SRY (sex determining region Y)-box 12 | Sox12 | 2010205A02Rik | 22.6kb | Anti-sense | Chr. 2 - 152393611 - 152398046 on NCBI Build GRCm38 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY512911.1, AY512942.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
